Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676251.3 | XP_005676308.1 | 3 - 4 | 1145 | 68 | |
XM_018061862.1 | XP_017917351.1 | 3 - 4 | 1104 | 68 | ||
XM_018061872.1 | XP_017917361.1 | 3 - 4 | 1260 | 68 | ||
XM_018061882.1 | XP_017917371.1 | 3 - 4 | 1111 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |