Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047386.1 | XP_017902875.1 | 2 - 3 | 443 | 66 | |
XM_018047387.1 | XP_017902876.1 | 3 - 4 | 720 | 66 | ||
XM_018047389.1 | XP_017902878.1 | 2 - 3 | 295 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |