Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684240.3 | XP_005684297.1 | 6 - 7 | 831 | 63 | |
XM_005684241.3 | XP_005684298.1 | 5 - 6 | 790 | 63 | ||
XM_013966237.2 | XP_013821691.1 | 6 - 7 | 793 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |