Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063947.1 | XP_017919436.1 | 2 - 3 | 646 | 76 | |
GenBank mRNA | JO334326.2 | - | 2 - 3 | 236 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |