Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSPG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018067113.1 | XP_017922602.1 | 3 - 4 | 2219 | 85 | |
GenBank mRNA | JO645739.2 | - | 3 - 4 | 1568 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |