Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962503.2 | XP_013817957.2 | 4 - 5 | 814 | 72 | |
XM_018045958.1 | XP_017901447.1 | 4 - 5 | 814 | 72 | ||
XM_018045959.1 | XP_017901448.1 | 4 - 5 | 814 | 72 | ||
XM_018045960.1 | XP_017901449.1 | 4 - 5 | 1059 | 72 | ||
GenBank mRNA | JO559054.2 | - | 4 - 5 | 826 | 72 | |
JO562932.2 | - | 4 - 5 | 826 | 72 | ||
JO586094.2 | - | 2 - 3 | 1616 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |