Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062063.1 | XP_017917552.1 | 2 - 3 | 369 | 63 | |
GenBank mRNA | JO633922.2 | - | 8 - 9 | 3385 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |