Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013967221.2 | XP_013822675.2 | 3 - 4 | 498 | 93 | |
XM_018053024.1 | XP_017908513.1 | 3 - 4 | 777 | 93 | ||
XM_018053030.1 | XP_017908519.1 | 3 - 4 | 576 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |