Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690387.3 | XP_005690444.1 | 2 - 3 | 220 | 93 | |
GenBank mRNA | JO412270.2 | - | 2 - 3 | 212 | 93 | |
JT698451.1 | - | 5 - 6 | 736 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |