Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UEVLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699526.3 | XP_005699583.1 | 5 - 6 | 599 | 85 | |
XM_005699529.3 | XP_005699586.1 | 2 - 3 | 218 | 85 | ||
XM_005699530.3 | XP_005699587.1 | 4 - 5 | 382 | 85 | ||
XM_013975842.2 | XP_013831296.1 | 5 - 6 | 599 | 85 | ||
XM_018043167.1 | XP_017898656.1 | 4 - 5 | 533 | 85 | ||
XM_018043168.1 | XP_017898657.1 | 5 - 6 | 599 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |