Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046441.1 | XP_017901930.1 | 2 - 3 | 384 | 93 | |
XM_018046479.1 | XP_017901968.1 | 2 - 3 | 384 | 93 | ||
XM_018046508.1 | XP_017901997.1 | 2 - 3 | 384 | 93 | ||
GenBank mRNA | JO339333.2 | - | 2 - 3 | 287 | 93 | |
JO339336.2 | - | 2 - 3 | 287 | 93 | ||
JO357678.2 | - | 1075 | 93 | |||
JO373282.2 | - | 2 - 3 | 198 | 93 | ||
JO422094.2 | - | 2 - 3 | 236 | 93 | ||
JO498976.2 | - | 2 - 3 | 241 | 93 | ||
JO556407.2 | - | 6 - 7 | 1147 | 93 | ||
JO579573.1 | - | 7 - 8 | 1097 | 93 | ||
JO634106.2 | - | 2 - 3 | 426 | 93 | ||
JT738573.1 | - | 351 | 93 | |||
KA306450.1 | - | 2 - 3 | 207 | 93 |
Gene Symbol: U2AF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO429735.2 | - | 1063 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |