Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVPR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047994.1 | XP_017903483.1 | 1 - 2 | 1382 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Capra hircus Probe spans exons |