Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFXAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013968267.2 | XP_013823721.2 | 2 - 3 | 1141 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons |