Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062404.1 | XP_017917893.1 | 20 - 21 | 9009 | 58 | |
XM_018062405.1 | XP_017917894.1 | 19 - 20 | 7761 | 58 | ||
GenBank mRNA | KA315085.1 | - | 19 - 20 | 6687 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |