Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060303.1 | XP_017915792.1 | 2 - 3 | 413 | 84 | |
XM_018060311.1 | XP_017915800.1 | 2 - 3 | 338 | 84 | ||
XM_018060315.1 | XP_017915804.1 | 3 - 4 | 452 | 84 | ||
XM_018060318.1 | XP_017915807.1 | 2 - 3 | 361 | 84 | ||
XM_018060324.1 | XP_017915813.1 | 2 - 3 | 414 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |