Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2H2orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676253.3 | XP_005676310.1 | 2 - 3 | 1067 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |