Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678196.3 | XP_005678253.2 | 7 - 8 | 1289 | 100 | |
XM_005678197.3 | XP_005678254.2 | 7 - 8 | 1289 | 100 | ||
XM_018045664.1 | XP_017901153.1 | 7 - 8 | 1289 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |