Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681043.3 | XP_005681100.3 | 4 - 5 | 830 | 92 | |
XM_005681045.3 | XP_005681102.2 | 3 - 4 | 524 | 92 | ||
GenBank mRNA | JR135256.1 | - | 4930 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |