Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681611.3 | XP_005681668.2 | 4 - 5 | 1431 | 82 | |
GenBank mRNA | JO608287.2 | - | 4 - 5 | 963 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |