Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681815.3 | XP_005681872.2 | 6 - 7 | 686 | 103 | |
XM_005681816.2 | XP_005681873.1 | 5 - 6 | 647 | 103 | ||
XM_005681817.2 | XP_005681874.1 | 5 - 6 | 658 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |