Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVPR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690409.2 | XP_005690466.2 | 1 - 2 | 942 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Capra hircus Probe spans exons |