Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692435.3 | XP_005692492.1 | 2 - 3 | 326 | 66 | |
XM_013971329.2 | XP_013826783.1 | 2 - 3 | 336 | 66 | ||
GenBank mRNA | JO441518.2 | - | 2 - 3 | 714 | 66 | |
JO616024.2 | - | 2 - 3 | 308 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |