Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692574.3 | XP_005692631.1 | 4 - 5 | 864 | 108 | |
XM_018062504.1 | XP_017917993.1 | 4 - 5 | 534 | 108 | ||
GenBank mRNA | JO452910.2 | - | 2 - 3 | 130 | 108 | |
KA335100.1 | - | 2 - 3 | 184 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |