Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963530.2 | XP_013818984.1 | 2 - 3 | 952 | 90 | |
GenBank mRNA | JO362898.2 | - | 1 - 2 | 479 | 90 | |
JO552716.2 | - | 1 - 2 | 246 | 90 | ||
JO597309.2 | - | 1 - 2 | 529 | 90 | ||
JO597310.2 | - | 1 - 2 | 529 | 90 | ||
JO647770.2 | - | 2 - 3 | 604 | 90 | ||
KA310525.1 | - | 2 - 3 | 528 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |