Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050376.1 | XP_017905865.1 | 3 - 4 | 509 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |