Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055489.1 | XP_017910978.1 | 3 - 4 | 1285 | 111 | |
GenBank mRNA | JO328752.2 | - | 3 - 4 | 516 | 111 | |
KA314323.1 | - | 3 - 4 | 1314 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |