Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNDIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013968621.2 | XP_013824075.1 | 3 - 4 | 1023 | 71 | |
XM_018057281.1 | XP_017912770.1 | 3 - 4 | 818 | 71 | ||
GenBank mRNA | JO644906.2 | - | 1 - 2 | 1135 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |