Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061773.1 | XP_017917262.1 | 4 - 5 | 661 | 83 | |
XM_018061774.1 | XP_017917263.1 | 4 - 5 | 595 | 83 | ||
GenBank mRNA | JO412150.2 | - | 4 - 5 | 573 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |