Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971288.2 | XP_013826742.2 | 11 - 12 | 1384 | 68 | |
XM_018062334.1 | XP_017917823.1 | 10 - 11 | 1471 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |