Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971288.2 | XP_013826742.2 | 25 - 26 | 3072 | 64 | |
XM_018062329.1 | XP_017917818.1 | 23 - 24 | 3439 | 64 | ||
XM_018062330.1 | XP_017917819.1 | 21 - 22 | 3315 | 64 | ||
XM_018062332.1 | XP_017917821.1 | 21 - 22 | 3312 | 64 | ||
XM_018062333.1 | XP_017917822.1 | 23 - 24 | 3285 | 64 | ||
XM_018062334.1 | XP_017917823.1 | 23 - 24 | 3267 | 64 | ||
XM_018062335.1 | XP_017917824.1 | 23 - 24 | 3216 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |