Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971288.2 | XP_013826742.2 | 6 - 7 | 845 | 68 | |
XM_018062329.1 | XP_017917818.1 | 6 - 7 | 999 | 68 | ||
XM_018062330.1 | XP_017917819.1 | 5 - 6 | 932 | 68 | ||
XM_018062332.1 | XP_017917821.1 | 5 - 6 | 932 | 68 | ||
XM_018062333.1 | XP_017917822.1 | 6 - 7 | 845 | 68 | ||
XM_018062334.1 | XP_017917823.1 | 5 - 6 | 932 | 68 | ||
XM_018062335.1 | XP_017917824.1 | 5 - 6 | 932 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |