Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20H5orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065515.1 | XP_017921004.1 | 3 - 4 | 832 | 101 | |
XM_018065516.1 | XP_017921005.1 | 3 - 4 | 720 | 101 | ||
XM_018065517.1 | XP_017921006.1 | 3 - 4 | 777 | 101 | ||
XM_018065518.1 | XP_017921007.1 | 3 - 4 | 665 | 101 | ||
GenBank mRNA | KA308590.1 | - | 2 - 3 | 434 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |