Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C26H10orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013975309.2 | XP_013830763.2 | 6 - 7 | 2327 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |