Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677179.3 | XP_005677236.1 | 2 - 3 | 212 | 83 | |
GenBank mRNA | JO452043.2 | - | 2 - 3 | 430 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |