Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049091.1 | XP_017904580.1 | 1 - 2 | 665 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |