Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680822.3 | XP_005680879.2 | 2 - 3 | 559 | 109 | |
GenBank mRNA | JO607296.2 | - | 2 - 3 | 302 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |