Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO599354.2 | - | 3 - 4 | 431 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |