Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692752.2 | XP_005692809.1 | 2 - 3 | 277 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |