Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039355.1 | XP_017894844.1 | 1 - 2 | 92 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |