Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061299.1 | XP_017916788.1 | 5 - 6 | 1243 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |