Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065431.1 | XP_017920920.1 | 4 - 5 | 548 | 59 | |
XM_018065432.1 | XP_017920921.1 | 4 - 5 | 1263 | 59 | ||
XM_018065433.1 | XP_017920922.1 | 4 - 5 | 796 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |