Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046285.1 | XP_017901774.1 | 7 - 8 | 872 | 61 | |
XM_018046286.1 | XP_017901775.1 | 7 - 8 | 1804 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Capra hircus Probe spans exons |