Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102188567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680860.3 | XP_005680917.2 | 4 - 5 | 555 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |