Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686286.3 | XP_005686343.1 | 4 - 5 | 853 | 79 | |
XM_005686287.3 | XP_005686344.1 | 4 - 5 | 835 | 79 | ||
XM_013967447.2 | XP_013822901.1 | 4 - 5 | 881 | 79 | ||
XM_018054914.1 | XP_017910403.1 | 4 - 5 | 785 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |