Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688299.3 | XP_005688356.1 | 3 - 4 | 313 | 107 | |
GenBank mRNA | JO338868.2 | - | 1 - 2 | 2644 | 107 | |
JO338870.2 | - | 1 - 2 | 2641 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |