Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698356.2 | XP_005698413.1 | 1 - 2 | 292 | 66 | |
GenBank mRNA | KA351780.1 | - | 1 - 2 | 330 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |