Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041983.1 | XP_017897472.1 | 7 - 8 | 1717 | 91 | |
XM_018041984.1 | XP_017897473.1 | 8 - 9 | 1758 | 91 | ||
GenBank mRNA | JI861797.1 | - | 6 - 7 | 796 | 91 | |
JO426877.1 | - | 7 - 8 | 1964 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |