Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699630.3 | XP_005699687.2 | 2 - 3 | 3238 | 93 | |
GenBank mRNA | JO510299.2 | - | 2 - 3 | 788 | 93 | |
KA336419.1 | - | 2 - 3 | 906 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |