Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054142.1 | XP_017909631.1 | 2 - 3 | 454 | 64 | |
GenBank mRNA | KA311078.1 | - | 2 - 3 | 81 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |