Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061796.1 | XP_017917285.1 | 3 - 4 | 1389 | 75 | |
XM_018061797.1 | XP_017917286.1 | 3 - 4 | 1280 | 75 | ||
XM_018061799.1 | XP_017917288.1 | 2 - 3 | 1291 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |